Learn More
Sino Biological Human NDRG3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 202006T10100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human NDRG3 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-3324