Learn More
ACROBiosystems Human Nectin-4-coupled Magnetic Beads
Catalog No. 502719175
Encompass_Preferred
Nectin-4(Nectin cell adhesion molecule 4) is a Nectin and nectin-like family of cell adhesion molecules. It is found in high levels in normal embryonic and fetal tissues, but decreases after adulthood and is limited in distribution in healthy tissues. Nectin-4 is overexpressed in a variety of tumor cells and promotes tumor cell growth and proliferation. Solid tumors with high expression of Nectin-4 include bladder cancer, pancreatic cancer, triple negative breast cancer, non-small cell lung cancer, stomach cancer, esophageal cancer, ovarian cancer and many other cancers. Studies have found that Nectin-4 promotes tumor cell proliferation, differentiation, migration and invasion by activating the PI3K/Akt pathway, and is believed to play an important role in the occurrence and metastasis of cancer. It is based on these research backgrounds that drug research targeting this target has emerged.
Catalog No. 50-271-9175 Supplier ACROBiosystems Supplier No. MBSK03910MG
Nectin-4(Nectin cell adhesion molecule 4) is a Nectin and nectin-like family of cell adhesion molecules. It is found in high levels in normal embryonic and fetal tissues, but decreases after adulthood and is limited in distribution in healthy tissues. Nectin-4 is overexpressed in a variety of tumor cells and promotes tumor cell growth and proliferation. Solid tumors with high expression of Nectin-4 include bladder cancer, pancreatic cancer, triple negative breast cancer, non-small cell lung cancer, stomach cancer, esophageal cancer, ovarian cancer and many other cancers. Studies have found that Nectin-4 promotes tumor cell proliferation, differentiation, migration and invasion by activating the PI3K/Akt pathway, and is believed to play an important role in the occurrence and metastasis of cancer. It is based on these research backgrounds that drug research targeting this target has emerged.