Learn More
ACROBiosystems Human Nectin-4 200ug
Catalog No. 502534386
Encompass_Preferred
Biotinylated Human Nectin-4 Protein FcAvitag (NE4-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).
Catalog No. 50-253-4386 Supplier ACROBiosystems Supplier No. NE4H82F4200UG
Biotinylated Human Nectin-4 Protein FcAvitag (NE4-H82F4) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).