Learn More
Sino Biological Anti-Nectin-1/PVRL1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11611MM09P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD111 / Nectin-1 / PVRL1 (rh CD111 / Nectin-1 / PVRL1; Catalog#11611-H08H; NP_002846.3; Met 1-Thr 334) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4667