Learn More
Sino Biological Anti-Nectin-1/PVRL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11611RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PVRL1 (rh PVRL1; Catalog#11611-H08H; NP_002846.3; Met 1-Thr 334). PVRL1 specific IgG was purified by human PVRL1 affinity chromatography.
Catalog No. 50-255-4666