Learn More
Sino Biological Recombinant Human Nectin-1/PVRL1 Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11611H02HB20
Encompass_Preferred
A DNA sequence encoding the human PVRL1 isoform 1 (NP_002846.3) extracellular domain (Met 1-Thr 334) was was fused with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4676