Learn More
Sino Biological Recombinant Anti-Nectin-2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10005R00520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD112 extracellular domain (rh CD112; Catalog#10005-H08H; Met 1-Leu 360; NP_002847.1).
Catalog No. 50-240-8099