Learn More
Sino Biological Human NECTIN3 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10852MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD113 / Nectin-3 / PVRL3 (rh CD113 / Nectin-3 / PVRL3; Catalog#10852-H08H; NP_056295.1; Met1-Asp400). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2616