Learn More
Sino Biological Recombinant Human Nectin-4 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 19771H02H1
Encompass_Preferred
A DNA sequence encoding the human NECTIN4 (NP_112178.2) (Met1-Ser349) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6403