Learn More
Biomatik Corporation Human NEDD4 Binding Protein 2 (N4BP2) ELISA Kit
Supplier:  Biomatik Corporation EKU09369
Encompass_Preferred
Human NEDD4 Binding Protein 2 (N4BP2) ELISA Kit for detecting N4BP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86UW6. Gene ID: 55728. Target Synonyms: B3BP; BCL3 Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7490