Learn More
Sino Biological Anti-COBRA1/NELFB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 206284T46100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human COBRA1 / NELFB fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-0272