Learn More
Biomatik Corporation Human Nephroblastoma Overexpressed Gene (NOV) ELISA Kit
Supplier:  Biomatik Corporation EKU11780
Encompass_Preferred
Human Nephroblastoma Overexpressed Gene (NOV) ELISA Kit for detecting NOV in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48745. Gene ID: 4856. Target Synonyms: CCN3; IGFBP9; NOVH; IBP-9; Insulin-like growth factor-binding protein 9; Protein NOV homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7472