Learn More
Raybiotech Inc Human Neprilysin ELISA Kit
Supplier:  Raybiotech Inc ELHNEPRILYSIN1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM-9, Angiopoietin 2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGF R, IFN-gammalpha, IGF-2, IGF-2 R, IL-1R6, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Notch-1, NOV, Osteoactivin, PD-1, PGRP-5, Serpin A4, sFRP-3, Thrombomodulin, TLR2, TRAIL R1, Transferrin, WIF-1ACE-2, Albumin, AMICA, Angiopoietin 4, BAFF, CA19-9, CD163 , Clusterin, CRTAM, CXCL14, Cystatin C, Decorin , Dkk-3, DLL1, Fetuin A, aFGF, FOLR1, Furin, GASP-1, GASP-2, GCSF R, HAI-2, IL-17B R, IL-27, LAG-3 , LDL R, Pepsinogen I, RANK, RBP4, SOST, Syndecan-1, TACI, TFPI, TSP-1, TRAIL R2 , TRANCE, Troponin I, uPA, VE-Cadherin, WISP-1
Catalog No. 50-194-9576