Learn More
American Research Products Inc Human NerVe growth factor NGF
Catalog No. 5000326373
Encompass
Human NerVe growth factor NGF ELISA kit from CUSABIO Detection Range 6 86 pg/ml-5000 pg/ml Sensitivity 6 86 pg/ml Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-26373 Supplier American Research Products Inc Supplier No. CSBE04683H
Human NerVe growth factor NGF ELISA kit from CUSABIO Detection Range 6 86 pg/ml-5000 pg/ml Sensitivity 6 86 pg/ml Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich