Learn More
Sino Biological Anti-Nestin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106970T44100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Nestin/NES, and purified by antigen affinity chromatography.
Catalog No. 50-240-8899