Learn More
Biomatik Corporation Human Nesprin 1 (Nesp1) ELISA Kit
Supplier:  Biomatik Corporation EKU06182
Encompass_Preferred
Human Nesprin 1 (Nesp1) ELISA Kit for detecting Nesp1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NF91. Target Synonyms: SYNE1; C6orf98; 8B; MYNE1; CPG2; SCAR8; ARCA1; Enaptin; Myocyte nuclear envelope protein 1; Nuclear envelope spectrin repeat protein 1; Synaptic nuclear envelope protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8846