Learn More
Biomatik Corporation Human Nesprin 2 (Nesp2) ELISA Kit
Supplier:  Biomatik Corporation EKU06183
Encompass_Preferred
Human Nesprin 2 (Nesp2) ELISA Kit for detecting Nesp2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8WXH0. Target Synonyms: SYNE2; NUA; NUANCE; SYNE-2; Synaptic nuclear envelope protein 2; Nuclear envelope spectrin repeat protein 2; Nucleus and actin connecting element protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7420