Learn More
BIOHIPPO Human NET1 Microsample
SDP
Catalog No. 5000313739
Encompass
Human NET1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NET1 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 132 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NET1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NET1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NET1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13739 Supplier BIOHIPPO Supplier No. BHE15211618
Human NET1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NET1 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 132 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NET1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NET1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NET1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color