Learn More
Biomatik Corporation Human N-Ethylmaleimide Sensitive Factor (NSF) ELISA Kit
Supplier:  Biomatik Corporation EKU06186
Encompass_Preferred
Human N-Ethylmaleimide Sensitive Factor (NSF) ELISA Kit for detecting NSF in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P46459. Target Synonyms: SKD2; N-Ethylmaleimide Sensitive Fusion Protein; Vesicle-fusing ATPase; Vesicular-fusion protein NSF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8845