Learn More
ACROBiosystems ACROBiosystems Human Netrin receptor DCC / DCC Protein, Fc Tag (MALS verified)
Catalog No. 502106359
Encompass_Preferred
Recombinant Protein;1MG;Human Netrin receptor DCC, Fc Tag (NEC-H5254) is expressed from human 293 cells (HEK293). It contains AA Phe 32 - Asn 1097 (Accession # P43146-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 144.1 kDa. The protein migrates as 140-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6359 Supplier ACROBiosystems Supplier No. NECH52541MG
Recombinant Protein;1MG;Human Netrin receptor DCC, Fc Tag (NEC-H5254) is expressed from human 293 cells (HEK293). It contains AA Phe 32 - Asn 1097 (Accession # P43146-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 144.1 kDa. The protein migrates as 140-160 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.