Learn More
ACROBiosystems Human Netrin receptor DCC / DCC Protein, His Tag
Catalog No. 502055259
Encompass_Preferred
Recombinant Protein;1MG;Human Netrin receptor DCC, His Tag (NEC-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 32 - Asn 1097 (Accession # P43146-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 119.5 kDa.
Catalog No. 50-205-5259 Supplier ACROBiosystems Supplier No. NECH52H31MG
Recombinant Protein;1MG;Human Netrin receptor DCC, His Tag (NEC-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 32 - Asn 1097 (Accession # P43146-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 119.5 kDa.