Learn More
Raybiotech Inc C-Series Human Neuro Discovery Array C1 Kit.
Catalog No. 501948625
Encompass_Preferred
Detects 20 human neurologically relevant proteins. Suitable for serum, plasma, cell culture media and other liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BDNF, beta-NGF, GCSF, GDNF, HB-EGF, IFN-gamma, IGF-1, IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-6, IL-8 (CXCL8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MMP-2, MMP-3, S100 B, TGF beta 1, TNF alpha, VEGF-A
Catalog No. 50-194-8625 Supplier Raybiotech Inc Supplier No. AAHNEU18
Detects 20 human neurologically relevant proteins. Suitable for serum, plasma, cell culture media and other liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: BDNF, beta-NGF, GCSF, GDNF, HB-EGF, IFN-gamma, IGF-1, IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-6, IL-8 (CXCL8), MCP-1 (CCL2), MIP-1 alpha (CCL3), MMP-2, MMP-3, S100 B, TGF beta 1, TNF alpha, VEGF-A