Learn More
Raybiotech Inc Quantibody Human Neuro Discovery Array 1 Kit.
Catalog No. 501950849
Encompass_Preferred
Detects 8 Human cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: BDNF, beta-NGF, CNTF, EGF, Eotaxin-1, Eotaxin-2, Eotaxin-3, Fas, GDNF, GM-CSF, IFN-gamma, IL-1 alpha, IL-1 beta, IL-10, IL-18, IL-4, IL-6, IL-8, LIF, MCP-1, MIP-1 beta, MMP-2, MMP-3, RANTES, TARC, TGF beta 1, TIMP-1, TNF alpha, TNF RI, VEGF-A
Catalog No. 50-195-0849 Supplier Raybiotech Inc Supplier No. QAHNEU11
Detects 8 Human cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: BDNF, beta-NGF, CNTF, EGF, Eotaxin-1, Eotaxin-2, Eotaxin-3, Fas, GDNF, GM-CSF, IFN-gamma, IL-1 alpha, IL-1 beta, IL-10, IL-18, IL-4, IL-6, IL-8, LIF, MCP-1, MIP-1 beta, MMP-2, MMP-3, RANTES, TARC, TGF beta 1, TIMP-1, TNF alpha, TNF RI, VEGF-A