Learn More
Biomatik Corporation Human Neuroblastoma, Suppression Of Tumorigenicity 1 (NBL1) CLIA Kit
Supplier:  Biomatik Corporation EKU08964
Encompass_Preferred
Human Neuroblastoma, Suppression Of Tumorigenicity 1 (NBL1) CLIA Kit for detecting NBL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P41271. Gene ID: 100532736. Target Synonyms: NB; DAN; NO3; DAND1; Differential Screening-Selected Gene Aberrant In Neuroblastoma; DAN domain family member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8583