Learn More
Biomatik Corporation Human Neuroblastoma, Suppression Of Tumorigenicity 1 (NBL1) ELISA Kit
Supplier:  Biomatik Corporation EKU06206
Encompass_Preferred
Human Neuroblastoma, Suppression Of Tumorigenicity 1 (NBL1) ELISA Kit for detecting NBL1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41271. Gene ID: 100532736. Target Synonyms: NB; DAN; NO3; DAND1; Differential Screening-Selected Gene Aberrant In Neuroblastoma; DAN domain family member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7082