Learn More
Biomatik Corporation Human Neurogenic Differentiation 6 (NEUROD6) ELISA Kit
Supplier:  Biomatik Corporation EKU06217
Encompass_Preferred
Human Neurogenic Differentiation 6 (NEUROD6) ELISA Kit for detecting NEUROD6 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96NK8. Gene ID: 63974. Target Synonyms: Atoh2; NEX1M; Math-2; bHLHa2; Neurogenic differentiation factor 6; Mammalian Atonal Homolog 2; Atonal Homolog 2; Class A basic helix-loop-helix protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7096