Learn More
Biomatik Corporation Human Neuromedin S (NMS) ELISA Kit
Supplier:  Biomatik Corporation EKU08702
Encompass_Preferred
Human Neuromedin S (NMS) ELISA Kit for detecting NMS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 123.5-10,000pg/ml. Sensitivity: 48.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q5H8A3. Gene ID: 129521. Target Synonyms: NM-S. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4487