Learn More
Biomatik Corporation Human Neuronal Apoptosis Inhibitory Protein (NAIP) ELISA Kit
Supplier:  Biomatik Corporation EKU06232
Encompass_Preferred
Human Neuronal Apoptosis Inhibitory Protein (NAIP) ELISA Kit for detecting NAIP in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13075. Gene ID: 4671. Target Synonyms: BIRC1; NLRB1; PsiNAIP; NLR Family,Apoptosis Inhibitory Protein; Baculoviral IAP Repeat-Containing Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8839