Learn More
Biomatik Corporation Human Neuropeptide FF (NPFF) CLIA Kit
Supplier:  Biomatik Corporation EKU11563
Encompass_Preferred
Human Neuropeptide FF (NPFF) CLIA Kit for detecting NPFF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.99pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: O15130. Gene ID: 8620. Target Synonyms: FMRFAL; Neuropeptide FF-Amide Peptide Precursor; Pro-FMRFamide-related neuropeptide FF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4716