Learn More
Sigma Aldrich Fine Chemicals Biosciences Human Neuropeptide Y NPY E
Catalog No. 502907882
Encompass
Neuropeptide Y (NPY) is a 36-amino acid peptide found primarily in the brain. It has been associated with a number of physiologic processes including the regulation of energy balance memory and learning. NPY is secreted by the hypothalamus and in addition to increasing appetite it increases the proportion of energy stored as fat and blocks signals to the brain. NPY is closely related to other peptide hormones such as pancreatic polypeptide PYY and correlates nicely to Leptin levels.
Catalog No. 50-290-7882 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. EZHNPY25K
Neuropeptide Y (NPY) is a 36-amino acid peptide found primarily in the brain. It has been associated with a number of physiologic processes including the regulation of energy balance memory and learning. NPY is secreted by the hypothalamus and in addition to increasing appetite it increases the proportion of energy stored as fat and blocks signals to the brain. NPY is closely related to other peptide hormones such as pancreatic polypeptide PYY and correlates nicely to Leptin levels.