Learn More
Biomatik Corporation Human Neuropeptide Y Receptor Y2 (NPY2R) ELISA Kit
Supplier:  Biomatik Corporation EKU06253
Encompass_Preferred
Human Neuropeptide Y Receptor Y2 (NPY2R) ELISA Kit for detecting NPY2R in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49146. Gene ID: 4887. Target Synonyms: NPY-Y2 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6739