Learn More
Biomatik Corporation Human Neuropeptide Y Receptor Y1 (NPY1R) ELISA Kit
Supplier:  Biomatik Corporation EKU06250
Encompass_Preferred
Human Neuropeptide Y Receptor Y1 (NPY1R) ELISA Kit for detecting NPY1R in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25929. Gene ID: 4886. Target Synonyms: NPYR; NPYY1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7578