Learn More
Raybiotech Inc Human/Mouse/Rat Neurophysin-2 EIA Kit.
Supplier:  Raybiotech Inc EIANEU21
Encompass_Preferred
Optimized for serum, plasma, and cell culture supernatants. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8826