Learn More
ACROBiosystems Human Neuropilin-1 / NRP1 / CD304 Protein, Mouse IgG2a Fc Tag (MALS verified)
Supplier:  ACROBiosystems NR1H52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human Neuropilin-1, Mouse IgG2a Fc Tag (NR1-H5253) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 96.7 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, His Tag (Cat. No. VE5-H5248) at 2 μg/mL (100 μL/well) can bind Human Neuropilin-1, Mouse IgG2a Fc Tag (Cat. No. NR1-H5253) with a linear range of 0.2-5 ng/mL (QC tested).
Catalog No. 50-205-4841