Learn More
Raybiotech Inc Human Neuropilin-2 ELISA Kit
Supplier:  Raybiotech Inc ELHNRP21
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 8.1 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Neuropilin-2.
Catalog No. 50-194-9597