Learn More
Biomatik Corporation Human Neuroplastin (NPTN) ELISA Kit
Supplier:  Biomatik Corporation EKU06263
Encompass_Preferred
Human Neuroplastin (NPTN) ELISA Kit for detecting NPTN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y639. Gene ID: 27020. Target Synonyms: SDR1; GP55; GP65; SDFR1; np55; np65; Stromal Cell Derived Factor Receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8834