Learn More
Raybiotech Inc Human/Mouse/Rat Neurotensin EIA Kit.
Supplier:  Raybiotech Inc EIANT1
Encompass_Preferred
Optimized for serum, plasma and cell culture supernatants. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8831