Learn More
Biomatik Corporation Human Neurotensin Receptor 2 (NTSR2) ELISA Kit
Supplier:  Biomatik Corporation EKU06269
Encompass_Preferred
Human Neurotensin Receptor 2 (NTSR2) ELISA Kit for detecting NTSR2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95665. Gene ID: 23620. Target Synonyms: NTR2; Levocabastine-sensitive neurotensin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7787