Learn More
Biomatik Corporation Human Neurotrophin 3 (NT3) CLIA Kit
Supplier:  Biomatik Corporation EKU08828
Encompass_Preferred
Human Neurotrophin 3 (NT3) CLIA Kit for detecting NT3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P20783. Gene ID: 4908. Target Synonyms: NTF3; HDNF; NGF2; Nerve Growth Factor 2; Neurotrophic Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4933