Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Neurotrypsin Synthetic Peptide

Catalog No. PEP0483 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0483 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0483 Supplier Invitrogen™ Supplier No. PEP0483
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

This peptide corresponds to 23 amino acids near the amino terminus of human Neurotrypsin PEP-0483 can be used as a blocking peptide with polyclonal antibody PA5-20363.

Neurotrypsin is a central nervous system-expressed serine protease whose truncation or absence causes nonsyndromic mental retardation. It is most prominently expressed in structures that are involved in the processing and storage of learned behavior and memory, such as the cerebral cortex, the hippocampus, and amygdala. Evidence suggests that neurotrypsin has multiple functions, including axonal outgrowth, maintaining neuronal plasticity, and arranging the perineuronal environment, partly in coordination with other proteases including tissue plasminogen activator. At least two isoforms of neurotrypsin are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 8492
Species Human
Purification Method purified
Accession Number P56730
Common Name Neurotrypsin
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias Brain-specific serine protease 3; Bssp3; BSSP-3; leydin; Motopsin; MRT1; Neurotrypsin; Nt; peptidase, serine, 12 (neurotrypsin, motopsin); protease, serine 12; protease, serine 12 neurotrypsin (motopsin); protease, serine, 12 (neurotrypsin, motopsin); protease, serine, 12 neurotrypsin (motopsin); Prss12; Serine protease 12
Gene Symbol PRSS12
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.