Learn More
Biomatik Corporation Human Neutral Sphingomyelinase Activation Associated Factor (NSMAF) ELISA Kit
Supplier:  Biomatik Corporation EKU08418
Encompass_Preferred
Human Neutral Sphingomyelinase Activation Associated Factor (NSMAF) ELISA Kit for detecting NSMAF in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92636. Gene ID: 8439. Target Synonyms: FAN; Factor associated with neutral sphingomyelinase activation. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8668