Learn More
Biomatik Corporation Human Neutral Sphingomyelinase (NSMASE) ELISA Kit
Supplier:  Biomatik Corporation EKU06277
Encompass_Preferred
Human Neutral Sphingomyelinase (NSMASE) ELISA Kit for detecting NSMASE in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60906. Gene ID: 6610. Target Synonyms: SMPD2; N-SMase; ISC1; Neutral Sphingomyelin Phosphodiesterase; Sphingomyelin Phosphodiesterase 2,Neutral Membrane; Lyso-platelet-activating factor-phospholipase C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6764