Learn More
American Research Products Inc Human neutrophil gelatinase as
Catalog No. 5000329812
Encompass
Human neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma urine tissue homogenates Method Sandwich
Catalog No. 50-003-29812 Supplier American Research Products Inc Supplier No. CSBE09408H
Human neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma urine tissue homogenates Method Sandwich