Learn More
BIOHIPPO Human NF-Bp105 Microsample
SDP
Catalog No. 5000314522
Encompass
Human NF-Bp105 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NF-Bp105 in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NF- Bp105 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NF- Bp105 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NF- Bp105 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14522 Supplier BIOHIPPO Supplier No. BHE15212084
Human NF-Bp105 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NF-Bp105 in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NF- Bp105 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NF- Bp105 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NF- Bp105 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color