Learn More
Sino Biological Anti-Neurofascin Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 15694MM02P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NFASC / Neurofascin (rh NFASC / Neurofascin; Catalog#15694-H08H; O94856-12; Met1-Gln939) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4086