Learn More
BIOHIPPO Human NFASC Microsample
SDP
Catalog No. 5000314371
Encompass
Human NFASC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NFASC in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NFASC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NFASC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NFASC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14371 Supplier BIOHIPPO Supplier No. BHE15211203
Human NFASC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NFASC in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NFASC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NFASC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NFASC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color