Learn More
ACROBiosystems Human NFL, His Tag (NFL-H5143) is expressed from E.coli cells. It contains AA Ser 2 - Asp 543 (Accession # P07196-1).
Supplier:  ACROBiosystems NFLH5143100UG
Encompass_Preferred
Neurofilaments (NF) are classed as type IV intermediate filaments found in the cytoplasm of neurons. Together with microtubules and microfilaments, they form the neuronal cytoskeleton. Neurofilaments usually contain three intermediate filament proteins: NFL, NFM, and NFH which are involved in the maintenance of neuronal caliber. Neurofilament heavy (NFH) can server as biomarkers for several diseases by indicating axonal injury.
Catalog No. 50-225-3912