Learn More
FUJIFILM BIOSCIENCES INC Human beta-NGF, Recombinant Protein, Shenandoah Biotechnology, 5 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 100385UG
Encompass_Preferred
Nerve growth factor beta (B-NGF) is a neurotrophic factor that is important for the development and maintenance of sensory and sympathetic neurons.  B-NGF signals through the low affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase A (TrkA) to activate PI3K, Ras, and PLC signaling pathways.  B-NGF is also involved in the growth, differentiation, and survival of B lymphocytes.  Human, mouse, and rat B-NGF proteins are cross-reactive. 
Catalog No. 50-990-926