Learn More
ACROBiosystems HEK293/Human NGFR Stable Cell Line
Catalog No. 502719743
Encompass_Preferred
The p75 neurotrophin receptor (p75NTR) was originally identified as a low-affinity receptor for neurotrophins(NGF R).p75NTR is a neurotrophic factor receptor. Neurotrophic factor receptors bind Neurotrophins including Nerve growth factor, Neurotrophin-3, Brain-derived neurotrophic factor, and Neurotrophin-4. All neurotrophins bind to p75NTR. This also includes the immature pro-neurotrophin forms. Neurotrophic factor receptors, including p75NTR, are responsible for ensuring a proper density to target ratio of developing neurons, refining broader maps in development into precise connections. p75NTR is involved in pathways that promote neuronal survival and neuronal death.
Catalog No. 50-271-9743 Supplier ACROBiosystems Supplier No. CHEKATP1572VIALS
The p75 neurotrophin receptor (p75NTR) was originally identified as a low-affinity receptor for neurotrophins(NGF R).p75NTR is a neurotrophic factor receptor. Neurotrophic factor receptors bind Neurotrophins including Nerve growth factor, Neurotrophin-3, Brain-derived neurotrophic factor, and Neurotrophin-4. All neurotrophins bind to p75NTR. This also includes the immature pro-neurotrophin forms. Neurotrophic factor receptors, including p75NTR, are responsible for ensuring a proper density to target ratio of developing neurons, refining broader maps in development into precise connections. p75NTR is involved in pathways that promote neuronal survival and neuronal death.