Learn More
BIOHIPPO Human NGN2 Microsample
SDP
Catalog No. 5000313823
Encompass
Human NGN2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NGN2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NGN2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NGN2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NGN2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13823 Supplier BIOHIPPO Supplier No. BHE15210583
Human NGN2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NGN2 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human NGN2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human NGN2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human NGN2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color